Läs mer
Piperacillin Methyl Ester, TRC
Produktkod. 30412475
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30412475 250 mg
30414978 500 mg
30412854 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30412475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP480065250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65700-41-6
Molekylformel C24 H29 N5 O7 S
Molekylvikt (g/mol) 531.58
Synonym Methyl (2S,5R,6R)-6-((R)-2-(4-Ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
IUPAC-namn methyl (2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
LEDER CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)OC)c4ccccc4)C(=O)C1=O
Specifikationer
Kemiskt namn eller material Piperacillin Methyl Ester
Kvantitet 250 mg
InChI formel InChI=1S/C24H29N5O7S/c1-5-27-11-12-28(20(33)19(27)32)23(35)26-14(13-9-7-6-8-10-13)17(30)25-15-18(31)29-16(22(34)36-4)24(2,3)37-21(15)29/h6-10,14-16,21H,5,11-12H2,1-4H3,(H,25,30)(H,26,35)/t14-,15-,16+,21-/m1/s1
Formel vikt 531.1788