Läs mer
Piperacillin Sodium, LoGiCal
Produktkod. 30460265
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30460265 10 mg
1 options
Produktkod. 30460265 Leverantör LoGiCal Leverantörsnummer LGCFOR0296.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Piperacillin Sodium
CAS 59703-84-3
Analyt- eller komponentnamn Piperacillin Sodium
Frakt skick Ice pack (-20°C)
Molekylformel C23 H26 N5 O7 S . Na
Certifikat och efterlevnad ISO 9001
Synonym Piperacillin Sodium, (2S,5R,6R)-6-[[(2R)-2-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt (1:1), (2S,5R,6R)-6-[[(2R)-2-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, CL 227193, Pentcillin, Piperacillin sodium, Piperacillin-Na, Sodium piperacillin, T 1220, [2S-[2alpha,5alpha,6beta(S*)]]-6-[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt
InChI formel InChI=1S/C23H27N5O7S.Na/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28;/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34);/q;+1/p-1/t13-,14-,15+,20-;/m1./s1
Lösningstyp Neat
LEDER [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)[O-])c4ccccc4)C(=O)C1=O
IUPAC-namn sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Industry Type Clinical Testing
Molekylvikt (g/mol) 539.54
Kvantitet 10 mg
Formel vikt 539.1451 g/mol