missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Piperacillin Sodium, Mikromol™
Description
Piperacillin Sodium
Specifications
Specifications
| Kemiskt namn eller material | Piperacillin Sodium |
| CAS | 59703-84-3 |
| Analyt- eller komponentnamn | Piperacillin Sodium |
| Frakt skick | Ice pack (-20°C) |
| Molekylformel | C23 H26 N5 O7 S . Na |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | Piperacillin Sodium, (2S,5R,6R)-6-[[(2R)-2-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium salt (1:1), (2S,5R,6R)-6-[[(2R)-2-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt, CL 227193, Pentcillin, Piperacillin sodium, Piperacillin-Na, Sodium piperacillin, T 1220, [2S-[2alpha,5alpha,6beta(S*)]]-6-[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid monosodium salt |
| InChI formel | InChI=1S/C23H27N5O7S.Na/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28;/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34);/q;+1/p-1/t13-,14-,15+,20-;/m1./s1 |
| LEDER | [Na+].CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)[O-])c4ccccc4)C(=O)C1=O |
| IUPAC-namn | sodium;(2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?