Läs mer
Piperazine Phosphate Monohydrate, TRC
Produktkod. 30419444
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30419444 10 g
1 options
Produktkod. 30419444 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP47988510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Piperazine Phosphate
CAS 14538-56-8
Molekylformel C4 H10 N2 . H3 O4 P
Kvantitet 10 g
Synonym Anthalazine Phosphate, Antipar, Parazine, Pincets, Pinrou, Pinsirup, Piperazate, Piperazine Monophosphate, Piperazine Phosphate
InChI formel InChI=1S/C4H10N2.H3O4P/c1-2-6-4-3-5-1;1-5(2,3)4/h5-6H,1-4H2;(H3,1,2,3,4)
LEDER OP(=O)(O)O.C1CNCCN1
IUPAC-namn phosphoric acid;piperazine
Molekylvikt (g/mol) 184.13
Formel vikt 184.0613