Läs mer
Piperazonifil, TRC
Produktkod. 30137407
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137407 100 mg
17983685 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137407 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP480470100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1335201-04-1
Molekylformel C25 H34 N6 O4
Molekylvikt (g/mol) 482.58
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[2-(4-ethyl-3-oxo-1-piperazinyl)-1-hydroxyethyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Piperazinonafil
IUPAC-namn 5-[2-ethoxy-5-[2-(4-ethyl-3-oxopiperazin-1-yl)-1-hydroxyethyl]phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(O)CN4CCN(CC)C(=O)C4
Specifikationer
Kemiskt namn eller material Piperazonifil
Kvantitet 100 mg
InChI formel InChI=1S/C25H34N6O4/c1-5-8-18-22-23(29(4)28-18)25(34)27-24(26-22)17-13-16(9-10-20(17)35-7-3)19(32)14-30-11-12-31(6-2)21(33)15-30/h9-10,13,19,32H,5-8,11-12,14-15H2,1-4H3,(H,26,27,34)
Formel vikt 482.2642