Läs mer
Piperidolate hydrochloride, MedChemExpress
Produktkod. 16481446
Encompass_Preferred
Change view
Kvantitet:
100 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16481446 200 mg
16458536 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16481446 Leverantör Medchem Express Leverantörsnummer HYB0962
MedChemExpress Piperidolate hydrochloride is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).
Piperidolate hydrochloride is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).
Specifikationer
Kemiskt namn eller material Piperidolate hydrochloride
Fysisk form Solid
CAS 129-77-1
Färg White
Kvantitet 200 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H26ClNO2
För användning med (applikation) Neuroscience-Neuromodulation
Löslighetsinformation H2O : 50 mg/mL (138.93 mM; Need ultrasonic) ∣DMSO : ≥ 32 mg/mL (88.92 mM)
LEDER O=C(OC1CN(CC)CCC1)C(C2=CC=CC=C2)C3=CC=CC=C3.[H]Cl
Molekylvikt (g/mol) 359.89
Formel vikt 359.89
Procent renhet 99.9%