missing translation for 'onlineSavingsMsg'
Läs mer

Piperidolate Hydrochloride, TRC

Produktkod. 30420430
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30420430 100 mg -
30410006 250 mg -
30415363 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30420430 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP479990100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 129-77-1
Molekylformel C21 H25 N O2 . Cl H
Molekylvikt (g/mol) 359.89
Synonym Benzeneacetic acid, α-phenyl-, 1-ethyl-3-piperidinyl ester, hydrochloride (1:1), Acetic acid, diphenyl-, 1-ethyl-3-piperidyl ester hydrochloride (6CI,7CI,8CI), Benzeneacetic acid, α-phenyl-, 1-ethyl-3-piperidinyl ester, hydrochloride (9CI), 3-Piperidinol, 1-ethyl-, diphenylacetate (ester), hydrochloride (8CI), Crapinon, Dactil hydrochloride, NSC 4349, Piperidolate hydrochloride
IUPAC-namn (1-ethylpiperidin-3-yl) 2,2-diphenylacetate;hydrochloride
LEDER Cl.CCN1CCCC(C1)OC(=O)C(c2ccccc2)c3ccccc3

Specifikationer

Kemiskt namn eller material Piperidolate Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C21H25NO2.ClH/c1-2-22-15-9-14-19(16-22)24-21(23)20(17-10-5-3-6-11-17)18-12-7-4-8-13-18;/h3-8,10-13,19-20H,2,9,14-16H2,1H3;1H
Formel vikt 359.1652
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.