missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.
Solubility
Soluble in water(0.04g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Piperine is used in increasing the bioavailability of some drugs and nutrients, it is used as medicine for Malaria and cholera, Stomach upset, Cancer.
Solubility
Soluble in water(0.04g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 128°C to 133°C |
Quantity | 5 g |
Beilstein | 90741 |
Merck Index | 14,7472 |
Solubility Information | Soluble in water(0.04g/L). |
Formula Weight | 285.34 |
Percent Purity | 98% |
Chemical Name or Material | Piperine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'