Läs mer
Piperoxan Hydrochloride, LoGiCal
Produktkod. 30462090
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30462090 5 mg
1 options
Produktkod. 30462090 Leverantör LoGiCal Leverantörsnummer LGCFOR1953.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Piperoxan Hydrochloride
CAS 135-87-5
Analyt- eller komponentnamn Piperoxan Hydrochloride
Frakt skick Room Temperature
Molekylformel C14 H19 N O2 . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Piperidine, 1-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, hydrochloride (1:1), Piperidine, 1-(1,4-benzodioxan-2-ylmethyl)-, hydrochloride (8CI), Piperidine, 1-[(2,3-dihydro-1,4-benzodioxin-2-yl)methyl]-, hydrochloride (9CI), Piperoxan hydrochloride (6CI), 1,4-Benzodioxin, piperidine deriv., 2-(1-Piperidylmethyl)-1,4-benzodioxan hydrochloride, 2-Piperidinomethyl-1,4-benzodioxan hydrochloride, 933F hydrochloride, Benodaine hydrochloride, Benzodioxane hydrochloride, Compd. 933F, F 933 hydrochloride, Fourneau 933 hydrochloride, Piperoxane hydrochloride, dl-Piperoxan hydrochloride
InChI formel InChI=1S/C14H19NO2.ClH/c1-4-8-15(9-5-1)10-12-11-16-13-6-2-3-7-14(13)17-12;/h2-3,6-7,12H,1,4-5,8-11H2;1H
Lösningstyp Neat
LEDER Cl.C(C1COc2ccccc2O1)N3CCCCC3
IUPAC-namn 1-(2,3-dihydro-1,4-benzodioxin-3-ylmethyl)piperidine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 269.77
Kvantitet 5 mg
Formel vikt 269.1183 g/mol