Läs mer
Piperoxan hydrochloride, MedChemExpress
Product Code. 16489736
Encompass_Preferred
Change view
Click to view available options
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
16489736 50 mg -
16483188 10 mg -
16410046 2 mg -
16405927 5 mg -
16404476 100 mg 100mg
16402666 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16489736 Supplier Medchem Express Supplier No. HY100850
MedChemExpress Piperoxan (Benodaine) hydrochloride is an α2 adrenoceptor antagonist. Piperoxan hydrochloride is the first-generation antihistamine.
Piperoxan (Benodaine) hydrochloride is an α2 adrenoceptor antagonist. Piperoxan hydrochloride is the first-generation antihistamine.
Specifications
Kemiskt namn eller material Piperoxan hydrochloride
CAS 135-87-5
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C14H20ClNO2
Kvantitet 50 mg
Synonym Benodaine hydrochloride
Löslighetsinformation H2O : 50 mg/mL (185.34 mM; Need ultrasonic) ∣DMSO : ≥ 31 mg/mL (114.91 mM)
LEDER [H]Cl.N1(CC2OC3=CC=CC=C3OC2)CCCCC1
Molekylvikt (g/mol) 269.77
Formel vikt 269.77
Procent renhet 99.66%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation