Learn More
Gilson™ PIPETMAN™ Comfort Handle
The ergonomically designed PIPETMAN™ Comfort Handle attaches to any PIPETMAN Classic, Neo, or G pipette to provide improved comfort and reduced pipetting fatigue.
Brand:  Gilson™ F161487
Additional Details : Weight : 0.25000kg
Product Code. 15787617
  • 10685.00 SEK / Pack of 50
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Large, more comfortable finger hook and ergonomically designed handle
  • Fits better in your hand
  • Offers better grip to support your daily pipetting activity and aids in reducing the risk of repetitive strain injuries
  • Contains an identification zone that allows you to mark the pipette using PIPETMAN™ Comfort Handle Labels
  • Easily attached to the pipette with no tools required
  • Labels are easily removed and the handle is autoclavable
  • White in color
  • Provided in packs of 50
Specifications
Specifications
Autoclavable
Ergonomic pipetting
PIPETMAN™ pipetting system
50/Pk.