Learn More
Microspec™ Pipette Tips
Available as sterile or non-sterile
Brand: Microspec™ AE5
Additional Details : Weight : 0.90000kg
Description
- Available as sterile or non-sterile
- In bulk bags or in grip seal bags
- Non-filtered
Specifications
Yes | |
Non-Filtered | |
Blue | |
Bulk Bag |
1000 μL | |
Non-sterile | |
Not Autoclavable | |
1000 |