Learn More
Brand™ Pipetting Aid
For BLAUBRAND™ intraEND micropipettes of 1uL or more.
Brand: Brand™ 709110
Additional Details : Weight : 0.80000kg
Description
- PET plastic tube
- Includes silicone adapter and TPE suction bellows with venting hole
- Provided in packs of 10
Specifications
Pipetting Aid | |
Blaubrand intraEND micropipettes >1ul | |
PET Plastic tube, silicone adapter and TPE suction bellows with venting hole | |
10/Pk. |