Learn More
BRAND™ Filter Tips
Free from DiHEMDA und Oleamid. Tips are manufactured under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used.
Brand:  BRAND™ 732508
Additional Details : Weight : 0.39000kg
 View more versions of this product
Product Code. 15154947
  • 585.00 SEK / Pack of 960
Estimated Shipment
12-03-2020
Add to basket
Description
Description
  • Cleanroom quality for contamination-free analyses
  • Perfect fit for the Transferpette™ micropipette and many other pipettes
  • Polypropylene construction
  • TipBox packaging
  • Non-sterile
  • Autoclavable at 121°C
  • DIN EN 285 and CE-marked according to IVD-Directive 98/79 EC
  • DNA-, RNase-, endotoxin-free
Specifications
Specifications
Bag
2 to 20μL
Non-sterile
Polypropylene
960/PK
Yes
Yes
DIN EN 285, CE-marked according to IVD-Directive 98/79 EC