Learn More
Brand™ Non-Sterile Polyethylene Filter Tips
Non-self-sealing filter tips from Brand™ have a PE filter that is free from chemical additives.
Brand:  Brand™ 732510
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 15164947
  • 655.00 SEK / Pack of 960
Estimated Shipment
09-03-2020
Add to basket
Description
Description
  • Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipet shaft
  • Functions with consistent reliability
  • Sample can be recovered from the filter by simply actuating the pipette’s blow-out function, or by centrifugation if necessary
  • Available in a wide range of capacities
Specifications
Specifications
Bulk: 1 bag of 960 each
Filter Tip
Yes
Yellow
Polyethylene
960
5 to 100µL
Non-sterile
Yes
Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween