Läs mer
Hirschmann™ Pipetus™ Standard Pipette Filler
Inkluderar:
Without filter on pump
Förpackningsstorlek:
Varje
Beskrivning
- Highly accurate pipetting enabled through fine dispensing valves with valve pins made of ceramic compound
- The seat of all pipettes is sealed absolutely tight with a silicone adapter
- Extremely light hand unit with external pump
- Hydrophobic PTFE diaphragm filter with 0.2 μm pore size
- Guaranteed precision, reliability and long service life
- With or without filter on pump
- Safety valve prevents the penetration of liquid into the interior of the unit
Specifikationer
Specifikationer
| Porstorlek | 0.2 μm |
| Typ | Pipette Filler |
| Volym (metrisk) | 0.1 to 200 mL |
| För användning med (applikation) | Pipetting |
| För användning med (utrustning) | Measuring, volumetric, Pasteur pipettes |
| Inkluderar | Without filter on pump |
| Pluggtyp | UK plug |
| Spänning | 230 V |