missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Piprinhydrinate, Mikromol™
Beskrivning
Piprinhydrinate
Specifikationer
Specifikationer
| Kemiskt namn eller material | Piprinhydrinate |
| CAS | 606-90-6 |
| Analyt- eller komponentnamn | Piprinhydrinate |
| Frakt skick | Room Temperature |
| Molekylformel | C19 H23 N O . C7 H7 Cl N4 O2 |
| Certifikat och efterlevnad | ISO 17034 |
| Synonym | Piprinhydrinate, 8-Chloro-3,9-dihydro-1,3-dimethyl-1H-purine-2,6-dione compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 8-Chloro-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 8-Chlorotheophylline compd. with 4-(diphenylmethoxy)-1-methylpiperidine (1:1), 4-(Diphenylmethoxy)-1-methylpiperidine compd. with 8-chloro-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione (1:1), 4-(Diphenylmethoxy)-1-methylpiperidine compd. with 8-chlorotheophylline (1:1), 8-Chlorotheophylline 4-(diphenylmethoxy)-1-methylpiperidine salt, Diphenylpyraline 8-chlorotheophyllinate, Diphenylpyraline theoclate, Kolton, Mepedyl, N-Methylpiperidyl 4-benzhydryl ether 8-chlorotheophyllinate |
| InChI formel | InChI=1S/C19H23NO.C7H7ClN4O2/c1-20-14-12-18(13-15-20)21-19(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h2-11,18-19H,12-15H2,1H3;1-2H3,(H,9,10) |
| LEDER | CN1CCC(CC1)OC(c2ccccc2)c3ccccc3.CN4C(=O)N(C)c5nc(Cl)[nH]c5C4=O |
| IUPAC-namn | 4-benzhydryloxy-1-methylpiperidine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione |
| Visa mer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?