Läs mer
Piracetam, TRC
Produktkod. 30012107
Change view
Kvantitet:
50 mg
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012107 50 g
30011193 50 mg
17949724 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012107 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP50080050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7491-74-9
Molekylformel C6 H10 N2 O2
Molekylvikt (g/mol) 142.16
Synonym Piracetam, (2-Oxopyrrolidino)acetamide, 2-(2-Oxopyrrolidin-1-yl)acetamide, 2-Ketopyrrolidine-1-acetamide, 2-Oxo-1-pyrrolidineacetamide, 2-Oxo-1-pyrrolidinylacetamide, 2-Pyrrolidinoneacetamide, 2-Pyrrolidoneacetamide, Avigilen, Axonyl, Cerebroforte, Cl 781, Encetrop, Euvifor, Gabacet, Genogris, Geram, Myocalm, Nootobril, Nootobryl, Nootron, Nootrop, Nootropil, Nootropyl, Normabrain, Norzetam, Piramem, Pirazetam, Pirroxil, Prima, Prima (pharmaceutical), Pyracetam, Pyramem, UCB 6215
IUPAC-namn 2-(2-oxopyrrolidin-1-yl)acetamide
LEDER NC(=O)CN1CCCC1=O
Specifikationer
Kemiskt namn eller material Piracetam
Kvantitet 50 g
InChI formel InChI=1S/C6H10N2O2/c7-5(9)4-8-3-1-2-6(8)10/h1-4H2,(H2,7,9)
Formel vikt 142.0742