Läs mer
Pirimicarb-desmethyl, TRC
Produktkod. 30424371
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424371 10 mg
30415219 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30424371 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP50876510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30614-22-3
Molekylformel C10 H16 N4 O2
Molekylvikt (g/mol) 224.26
Synonym Carbamic acid, dimethyl-, 5,6-dimethyl-2-(methylamino)-4-pyrimidinyl ester (8CI,9CI), 5,6-Dimethyl-2-(methylamino)-4-pyrimidinyl dimethylcarbamate, Pirimicarb III, Pirimicarb-desmethyl, R34836
IUPAC-namn [5,6-dimethyl-2-(methylamino)pyrimidin-4-yl] N,N-dimethylcarbamate
LEDER CNc1nc(C)c(C)c(OC(=O)N(C)C)n1
Specifikationer
Kemiskt namn eller material Pirimicarb-desmethyl
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C10H16N4O2/c1-6-7(2)12-9(11-3)13-8(6)16-10(15)14(4)5/h1-5H3,(H,11,12,13)
Formel vikt 224.1273
Procent renhet >95%