Läs mer
Pirimicarb, TRC
Produktkod. 30426715
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30426715 1 g
1 options
Produktkod. 30426715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP5087681G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pirimicarb
CAS 23103-98-2
Molekylformel C11 H18 N4 O2
Kvantitet 1 g
Synonym Carbamic acid','dimethyl-','2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester (8CI,9CI), 4-Pyrimidinol','2-(dimethylamino)-5,6-dimethyl-','dimethylcarbamate (ester) (8CI), 2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethylcarbamate, 2-Dimethylamino-5,6-dimethylpyrimidin-4-yl N,N-dimethylcarbamate, 5,6-Dimethyl-2-dimethylamino-4-dimethylcarbamoyloxypyrimidine, Aficida, Aphox, Aphox DG, Fernos, PP 062, Patton Flow, Pirimicarb, Pirimix, Pirimor, Pirimor G, Pirimor Granulate, Pyrimor, Carbamic acid','N,N-dimethyl-','2-(dimethylamino)-5,6-dimethyl-4-pyrimidinyl ester
InChI formel InChI=1S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3
LEDER CN(C)C(=O)Oc1nc(nc(C)c1C)N(C)C
IUPAC-namn [2-(dimethylamino)-5,6-dimethylpyrimidin-4-yl] N,N-dimethylcarbamate
Molekylvikt (g/mol) 238.29
Formel vikt 238.143