Läs mer
Pitavastatin Calcium, MedChemExpress
Produktkod. 16498358
Encompass_Preferred
Change view
Kvantitet:
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16498358 10 mg -
16424257 50 mg -
16408136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16498358 Leverantör Medchem Express Leverantörsnummer HYB0144
MedChemExpress Pitavastatin Calcium (NK-104 hemicalcium) is a potent hydroxymethylglutaryl-CoA (HMG-CoA) reductase inhibitor.
Pitavastatin Calcium (NK-104 hemicalcium) inhibits cholesterol synthesis from acetic acid with an IC50 of 5.8 nM in HepG2 cells. Pitavastatin Calcium is an efficient hepatocyte low-density lipoprotein-cholesterol (LDL-C) receptor inducer. Pitavastatin Calcium also possesses anti-atherosclerotic, anti-asthmatic, anti-osteoarthritis, antineoplastic, neuroprotective, hepatoprotective and reno-protective effects.
Specifikationer
Kemiskt namn eller material Pitavastatin Calcium
CAS 147526-32-7
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C25H23FNO4.1/2Ca
Kvantitet 10 mg
Synonym NK-104 hemicalciumPitavastatin hemicalcium
Löslighetsinformation DMSO : ≥ 50 mg/mL (113.51 mM)
LEDER O=C([O-])C[C@H](O)C[C@H](O)/C=C/C1=C(C2=CC=C(F)C=C2)C3=CC=CC=C3N=C1C4CC4.[0.5Ca2+]
Molekylvikt (g/mol) 440.49
Formel vikt 440.49
Procent renhet 97.0%
Kvalitet Research
Fysisk form Powder
För användning med (applikation) Cancer-programmed cell death