Läs mer
Pitavastatin Methyl Ester, TRC
Produktkod. 30415857
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30415857 250 mg
1 options
Produktkod. 30415857 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP531015250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pitavastatin Methyl Ester
CAS 849811-78-5
Molekylformel C26 H26 F N O4
Kvantitet 250 mg
Synonym Pitavastatin Methyl Ester, (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Methyl Ester, Methyl (3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoate, DOLM, Pitavastatin Methyl Ester
InChI formel InChI=1S/C26H26FNO4/c1-32-24(31)15-20(30)14-19(29)12-13-22-25(16-8-10-18(27)11-9-16)21-4-2-3-5-23(21)28-26(22)17-6-7-17/h2-5,8-13,17,19-20,29-30H,6-7,14-15H2,1H3/b13-12+/t19-,20-/m1/s1
LEDER COC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4
IUPAC-namn methyl (E,3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate
Molekylvikt (g/mol) 435.487
Formel vikt 435.185