Läs mer
Pitavastatin (+)-Phenylethylamine, TRC
Produktkod. 30420450
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30420450 100 mg
1 options
Produktkod. 30420450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR272515100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pitavastatin (+)-Phenylethylamine
CAS 147511-70-4
Molekylformel C25H24FNO4 • C8H11N
Kvantitet 100 mg
Synonym (3R,5S)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolyl]-3,5-dihydroxy-6-heptane Acid (+)-Phenylethylamine Salt, (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid (αR)-α-Methylbenzenemethanamine (1:1), [S-[R*,S*-(E)]]-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid (αR)-α-Methylbenzenemethanamine (1:1), (R)-α-Methylbenzenemethanamine [S-[R*,S*-(E)]]-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoate, (αR)-α-Methylbenzenemethanamine (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoate
InChI formel InChI=1S/C25H24FNO4.C8H11N/c26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31;1-7(9)8-5-3-2-4-6-8/h1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31);2-7H,9H2,1H3/b12-11+;/t18-,19-;7-/m11/s1
LEDER C[C@H](C1=CC=CC=C1)N.O[C@@H](C[C@@H](O)CC(O)=O)/C=C/C2=C(C3=CC=C(F)C=C3)C(C=CC=C4)=C4N=C2C5CC5
IUPAC-namn (R)-1-phenylethan-1-amine (3R,5S,E)-7-(2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl)-3,5-dihydroxyhept-6-enoate
Molekylvikt (g/mol) 542.64
Formel vikt 542.26