Learn More
Pivalic Acid, Sigma-Aldrich™
Product Code. 15888520
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888520 1 L 1 L
15878520 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15888520 Supplier Sigma Aldrich Supplier No. 8013361000
 Restricted Item
Beräknad leverans: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-98-9
Specifications
Melting Point 32°C to 35°C
Density 0.91 g/cm3 at 20°C
Molecular Formula (CH3)3CCOOH
MDL Number MFCD00004194
Quantity 1 L
Synonym Pivalic acid, Neopentanoic acid, 2,2-Dimethylpropionic acid, Trimethylacetic acid, Pivalic acid
Molecular Weight (g/mol) 102.13
Grade Research