Learn More
Honeywell Safety Products™ Pivot Silver/Black, Clear FogBan Lens Glasses
Click to view available options
Frame Color:
Silver, Black
Description
- General use and protection against impact (45m/s) during mechanical work such as grinding, turning and milling.
- Protection against impact from −5°C to +55°C.
- Sport styling and cushioned comfort.
- Two-tone, multi-material frame.
- Wraparound 8-base lens provides exceptional coverage.
- Extra cheek and side coverage.
- Quick and easy lens replacement system.
- Lightweight profile.
- Ratcheting temple angle adjustment.
- Ventilation channels minimize fogging.
- Elastomer nasal pads minimize slippage.
Specifications
Certifications/Compliance | EC Category PPE 2, EC Certificate Number EC1255, EC Attestation Number EC1255 |
Lens Coating | Anti-fog, Impact-resistant |
Product Type | Safety glasses |
For Use With (Application) | Administration, General use and protection against impact (45m/s), Mechanical Work (grinding, turning, milling), Protection against impact from −5°C to +55°C |
Frame Color | Silver, Black |
No. per Pack | 1 Ea. |
Weight (Metric) | 21 g |
Lens Material | Polycarbonate |
Frame Material | Plastic |