Läs mer
Pizotifen Malate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Pizotifen Malate |
| CAS | 5189-11-7 |
| Molekylformel | C19 H21 N S . C4 H6 O5 |
| Kvantitet | 100 mg |
| Synonym | Butanedioic acid','2-hydroxy-','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1), Butanedioic acid','hydroxy-','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1) (9CI), Malic acid','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (1:1) (8CI), Malic acid','compd. with 4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidine (7CI), 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene','9,10-dihydro-4-(1-methyl-4-piperidylidene)-','malate (1:1) (8CI), 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene','piperidine deriv., Piperidine','4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-','hydroxybutanedioate (1:1) (9CI), Piperidine','4-(9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methyl-','malate (1:1) (8CI), 4-(9,10-Dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thien-4-ylidene)-1-methylpiperidinium hydrogen malate, Mosegor, Pizotifen hydrogen malate, Pizotifen malate, Pizotyline malate, Sanmigran, Sanomigran |
| InChI formel | InChI=1S/C19H21NS.C4H6O5/c1-20-11-8-15(9-12-20)19-16-5-3-2-4-14(16)6-7-18-17(19)10-13-21-18;5-2(4(8)9)1-3(6)7/h2-5,10,13H,6-9,11-12H2,1H3;2,5H,1H2,(H,6,7)(H,8,9) |
| LEDER | CN1CCC(=C2c3ccccc3CCc4sccc24)CC1.OC(CC(=O)O)C(=O)O |
| IUPAC-namn | 4-(4,5-dihydrobenzo[1,2]cyclohepta[2,4-b]thiophen-10-ylidene)-1-methylpiperidine;2-hydroxybutanedioic acid |
| Molekylvikt (g/mol) | 429.53 |
| Formel vikt | 429.161 |