missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
PKC alpha Antibody (G13-R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP1-30121-0.05ML
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PKC alpha Monoclonal specifically detects PKC alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifikationer
| PKC alpha | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:5000, ELISA 1:20000-1:50000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P17252 | |
| PRKCA | |
| Peptide derived from the human PKC-alpha sequence | |
| 0.05 mL | |
| Cancer, Cellular Markers, mTOR Pathway, Signal Transduction, Wnt Signaling Pathway | |
| 5578 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| G13-R | |
| Unconjugated | |
| 20mM Tris/HCl (pH 8.0) and 10 mg/ml BSA with 0.05% Sodium Azide | |
| AAG6, aging-associated gene 6, EC 2.7.11, EC 2.7.11.13, MGC129901, PKC-A, PKC-alpha, PKCAMGC129900, PRKACA, protein kinase C alpha type, protein kinase C, alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering