Learn More
Invitrogen™ PKN1 (PRK1) Recombinant Human Protein
PKN1 (PRK1) belongs to the protein kinase C superfamily.
Brand: Invitrogen™ PV4037
Additional Details : Weight : 0.00900kg
Description
PKN1 (PRK1) is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids.
Specifications
NP_998725 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
1mg | |
RUO - research use only | |
PKN1 (PRK1) | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
132.1kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
PKN Kinase Family | |
Recombinant | |
Baculovirus |