missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ PKP3 Polyclonal Antibody

Beskrivning
Antibody detects endogenous levels of total PKP3.
Plakophilin-3 is an Armadillo-like protein present in nuclei and desmosomes of epithelial cells. The expression pattern of this protein seems to be largely restricted to epithelial cell types. Plakophilin-3 can be detected along cell borders in a punctuate staining pattern typical for desmosomal proteins. In addition to the desmosomal immunolocalization, immunostaining was observed as bright nuclear speckles. Thus, like plakophilin-1 and-2, plakophilin-3 displays a dual intracellular localization in the desmosomal plaque and in the cell nucleus, and therefore is probably involved in signal transduction pathways between the plasma membrane and the nucleus. The human protein has a predicted molecular mass of ~87 kDa.
Specifikationer
Specifikationer
| Antigen | PKP3 |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gen | PKP3 |
| Genaccessionsnr. | Q9QY23, Q9Y446 |
| Gene Alias | 2310056L12Rik; Pkp3; plakophilin 3; plakophilin 3b; plakophilin-3 |
| Gensymboler | PKP3 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?