missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PLA2G4B Recombinant Protein

Brand:  Abnova™ H00008681-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16172762

  • 4515.00 SEK / 10µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH25290.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
60.9
8
Glutathione Sepharose 4 Fast Flow
10 ug
MAEAALEAVRSELREFPAAARELCVPLAVPYLDKPPTPLHFYRDWVCPNRPCIIRNALQHWPALQKWSLPYFRATVGSTEVSVAVTPDGYADAVRGDRFMMPAERRLPLSFVLDVLEGRAQHPGVLYVQKQCSNLPSELPQLLPDLESHVPWASEALGKMPDAVNFWLGEAAAVTSLHKDHYENLYCVVSGEKHFLFHPPSDRPFIPYELYTPATYQLTEEGTFKVVDEEAMEKVPWIPLDPLAPDLARYPSYSQAQALRCTVRAGEMLYLPALWFHHVQQSQGCIAVNFWYDMEYDLKYSYFQLLDSLTKASGLD
HsT16992/PLA2G4B/cPLA2-beta
JMJD7-PLA2G4B
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
8681
PLA2G4B (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
JMJD7-PLA2G4B
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers