Learn More
PLA2R1 Monoclonal Antibody (CL0485), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524608
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%. Binds to an epitope located within the peptide sequence VKNCEERLFYICKKA as determined by overlapping synthetic peptides.
PLA2R1 Monoclonal antibody specifically detects PLA2R1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
PLA2R1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13018 | |
Pla2r1 | |
Recombinant protein corresponding to Human PLA2R1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL0485 | |
Unconjugated | |
Pla2r1 | |
180 kDa secretory phospholipase A2 receptor; CLEC13C; C-type lectin domain family 13 member C; mannose receptor, C type 2; Mrc2; M-type receptor; phospholipase A2 receptor 1; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; phospholipase A2, group IB, pancreas, receptor; Pla2g1br; PLA2G1R; PLA2-I receptor; PLA2IR; Pla2r; Pla2-r; PLA2R1; Secretory phospholipase A2 receptor; Soluble PLA2-R; Soluble secretory phospholipase A2 receptor | |
Mouse | |
Protein A | |
RUO | |
22925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only