missing translation for 'onlineSavingsMsg'
Learn More

PLAA Mouse anti-Human, Mouse, Rat, Clone: 5C10G9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60244-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887275

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Phospholipase A-2-activating protein (PLAA), also named PLAP, can interact with ubiquitin and is involved in the maintece of ubiquitin levels. PLAA contains multiple consensus domains including WD repeats, PFU domain and PUL domain. Although the function of PLAA remains unclear, ubiquitin binding of PLAA might be the central role that connects ubiquitination and degradation in ERAD. Recent finding revealed that PLAA served as a novel nociceptive mediator after incision, and its expression level is regulated by miR-203.
Specifications

Specifications

PLAA
5C10G9
Unconjugated
PLAA
Liquid
Plaa
PLAA Fusion Protein Ag3224
Protein A
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P27612, P54319, Q9Y263
DOA1, PLA2P, PLAA, PLAP
Mouse
IgG1
20 μL
-20°C
Primary
116645, 18786, 9373
Videos
SDS
Documents
Certificates
Special Offers

Special Offers