Läs mer
Plasmid DNA isolation solution III, Thermo Scientific™
Kvantitet:
250 ml
500 ml
Förpackningsstorlek:
250ml
500ml
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
This solution returns the pH to neutral after lysis with SDS and NaOH and allows DNA strands to renature and partially rehybridize. At the same time, the potassium acetate precipitates the detergent from the cell suspension.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Plasmid DNA isolation solution III |
| Kvantitet | 250 ml |
| Fysisk form | Liquid |