Learn More
RETSCH Plastic Collecting Receptacles
Brand:  RETSCH 22.003.0010
54150.00 SEK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 3.00000kg
 View more versions of this product
Product Code. 11964332
  • 57000.00 SEK / Each
Estimated Shipment: 03-07-2024
to see stock.
Specifications
Collecting Receptacle
For Cutting Mill SM 200
Plastic
30 L
Adapter and Filter Hose