Learn More
Multiroir Controlec Plastic Stackable Storage Box
Product Code. 16477070
Change view
Click to view available options
Capacity (Metric):
53 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
16477070 53 L Each
1 options
Product Code. 16477070 Supplier Multiroir Controlec Supplier No. 187072780
  • ONLINE EXCLUSIVE PRICE
    408.04 SEK
    257.13 SEK / Paket med 1
    Save 150.91 SEK
    36% Av
I lager
Add to basket
Add to basket
The Multiroir stackable storage boxes are suitable for food use and feature ergonomic handles for easy transporation.
  • Resistant to temperature variations
  • Stackable and nestable
  • Insensitive to shocks
  • Ergonomic handles for easy transport
  • Food contact quality
Specifications
Capacity (Metric) 53 L