Learn More
Karl Hecht™ Plastic Stirring Spatula
Designed as a stirrer for rectangular cuvettes. Karl Hecht™ Plastic Stirring Spatula is T-shaped with a cross-bar for stirring.
Brand:  Karl Hecht™ 40734010
Additional Details : Weight : 0.10000kg
Product Code. 10757312
  • 278.78 SEK / Pack of 100
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
  • T-shaped.
  • Plastic construction.
  • For rectangular cuvettes.
Specifications
Specifications
100 Pk.
Rectangular cuvettes
Plastic
Stirring Spatula
9 mm
120 mm