Learn More
Kartell™ Plastilab™ Untapered T-Shaped Tubing Connectors
Designed with serrations to hold hoses tightly and prevent slipping.
Brand:  Kartell™ 7046000
Additional Details : Weight : 0.00900kg
Product Code. 11994349
  • 79.60 SEK / Pack of 10
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Description
Description
Designed with serrations to hold hoses tightly and prevent slipping.
  • Made from polypropylene
  • Autoclavable
  • Chemical- and temperature-resistant
Compliance
Meet Food and Agriculture regulations