Learn More
Eppendorf™ Platform for Innova™ 2050 Shaker
This dedicated platform fits shaker model 2050.
Brand:  Eppendorf™ M1190-9903
 View more versions of this product
Product Code. 11628623
  • 8760.00 SEK / Each
In Stock
to see stock.
Explore more special offers
Description
  • Holds Erlenmeyer flasks
  • Dedicated platform
  • Made of stainless steel
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL Erlenmeyer Flasks
41 x 31 cm
35
Stainless Steel
Innova™ 2000 Shaker