missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Platinum, 10% on activated carbon paste, Type

Brand:  Alfa Aesar™ H36044.14

Additional Details : CAS Number : 7440-06-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15449065

  • 4934.52 SEK / 25g


EU Stock 3
Add to basket

Description

Description

Applications
Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Insoluble in water, mineral acids, and organic acids. Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, organic materials and flammable substances.
Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

Platinum
7440-06-4
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Solid
Black
10% on activated carbon paste, Type 128M
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
Odorless
25g
Videos
SDS