missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 10% on activated carbon paste, Type 128M, Thermo Scientific Chemicals

Catalog Number 15497607
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum as a catalyst is used for hydrogenation of alkenes, alkynes, nitro groups. It is also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Insoluble in water, mineral acids, and organic acids. Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, organic materials and flammable substances.
TRUSTED_SUSTAINABILITY
Color Black
Physical Form Solid
Quantity 5 g
Solubility Information Insoluble in water,mineral acids,and organic acids. Soluble in boiling aqua regia (reacts),and molten alkali cyanides (reacts).
Odor Odorless
Chemical Name or Material Platinum, 10% on activated carbon paste, Type 128M

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'