missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on graphite, Type 287, Thermo Scientific Chemicals

Catalog Number 15432917
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Physical Form Powder
Quantity 5 g
UN Number UN1325
Odor Odorless
Chemical Name or Material Platinum, 5% on graphite, Type 287

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'