Learn More
Platinum 6 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
8591.31 SEK - 28767.87 SEK
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11340168
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042914.BU
25 cm
8591.31 SEK
25cm
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
11350168
SDS View Documents Promotion Details
Thermo Scientific Alfa Aesar
042914.BY
100 cm
28767.87 SEK
100cm
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Description
Description
Platinum rhodium wire is used as a catalyst in preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.