missing translation for 'onlineSavingsMsg'
Learn More

Platinum, nominally 70% on high surface area advanced carbon support, HiSPEC™ 13100, Thermo Scientific™

Catalog Number p-7059633
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
23939
CHEBI:33400
platinum
[Pt]
This item is not returnable. View return policy
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
23939
CHEBI:33400
platinum
[Pt]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Gray to Black
Odor Odorless
Quantity 1 g
UN Number UN1325
Solubility Information Insoluble in water.
Chemical Name or Material Platinum, HiSPEC™ 13100, Nominally 70% on high surface area advanced carbon support
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'