Läs mer
Platonin Diiodide, TRC
Produktkod. 30422296
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422296 10 mg
30418514 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422296 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP57821010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3571-88-8
Molekylformel C38H61I2N3S3
Molekylvikt (g/mol) 909.91
Synonym 4,4'-Dimethyl-3,3'-di-n-heptyl-8-[2-(4-methyl-3-n-heptylthiazole)]2,2'-dicarbocyanin diiodide, 4,4'-Dimethyl-3,3'-dihepthyl-8-[2-(4-methyl-3-heptylthiazole)]-2,2'-dicarbocyanine diiodide, 4,4'-Dimethyl-3,3'-diheptyl-8-[2-(4-methyl-3-heptylthiazole)]-2,2'-dicarbocyanine diiodide, Kankohso 101, NK 19, Photosensitizer 101, Platonin J, Platonin Diiodide, Platonin Iodide, 2,2'-[3-[2-(3-Heptyl-4-methyl-4-thiazolin-2-ylidene)ethylidene]propenylene]bis[3-heptyl-4-methyl-thiazolium Diiodide
IUPAC-namn 2-[3,5-bis(3-heptyl-4-methyl-1,3-thiazol-3-ium-2-yl)penta-2,4-dienylidene]-3-heptyl-4-methyl-1,3-thiazole;diiodide
LEDER [I-].[I-].CCCCCCCN1C(=CSC1=CC=C(C=Cc2scc(C)[n+]2CCCCCCC)c3scc(C)[n+]3CCCCCCC)C
Specifikationer
Kemiskt namn eller material Platonin Diiodide
Kvantitet 10 mg
InChI formel InChI=1S/C38H61N3S3.2HI/c1-7-10-13-16-19-26-39-32(4)29-42-36(39)24-22-35(38-41(34(6)31-44-38)28-21-18-15-12-9-3)23-25-37-40(33(5)30-43-37)27-20-17-14-11-8-2;;/h22-25,29-31H,7-21,26-28H2,1-6H3;2*1H/q+2;;/p-2
Formel vikt 909.2117