Läs mer
Plazomicin Sulfate (>85%), TRC
Produktkod. 30422724
Change view
Kvantitet:
0.25 mg
1 mg
2.5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422724 0.25 mg
30416545 2.5 mg
30411904 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30422724 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP5795050.25MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1380078-95-4
Molekylformel C25H48N6O10 . x (H2SO4)
Molekylvikt (g/mol) 690.8
Synonym O-2-Amino-2,3,4,6-tetradeoxy-6-[(2-hydroxyethyl)amino]-alpha-D-glycero-hex-4-enopyranosyl-(1-4)-O-[3-deoxy-4-C-methyl-3-(methylamino)-beta-L-arabinopyranosyl-(1-6)]-N1-[(2S)-4-amino-2-hydroxy-1-oxobutyl]-2-deoxy-D-streptamine Sulphate Salt
IUPAC-namn (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-4-[[(2S,3R)-3-amino-6-[(2-hydroxyethylamino)methyl]-3,4-dihydro-2H-pyran-2-yl]oxy]-2-[(2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)oxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide;sulfuric acid
LEDER CN[C@@H]1[C@@H](O)[C@@H](O[C@@H]2[C@@H](O)[C@H](O[C@H]3OC(=CC[C@H]3N)CNCCO)[C@@H](N)C[C@H]2NC(=O)[C@@H](O)CCN)OC[C@]1(C)O.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Plazomicin Sulfate
Kvantitet 0.25 mg
InChI formel InChI=1S/C25H48N6O10.H2O4S/c1-25(37)11-38-24(18(35)21(25)29-2)41-20-15(31-22(36)16(33)5-6-26)9-14(28)19(17(20)34)40-23-13(27)4-3-12(39-23)10-30-7-8-32;1-5(2,3)4/h3,13-21,23-24,29-30,32-35,37H,4-11,26-28H2,1-2H3,(H,31,36);(H2,1,2,3,4)/t13-,14+,15-,16+,17+,18-,19-,20+,21-,23-,24-,25+;/m1./s1
Formel vikt 690.3106