missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26830-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Binds specifically to phosphatidylinositol 3,4-diphosphate (PtdIns3,4P2), but not to other phosphoinositides. May recruit other proteins to the plasma membrane.Specifications
PLEKHA1 | |
Polyclonal | |
Unconjugated | |
PLEKHA1 | |
PLEKHA1, TAPP 1, TAPP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
59338 | |
-20°C | |
Liquid |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9HB21 | |
PLEKHA1 | |
PLEKHA1 Fusion Protein Ag25285 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |