missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLEKHA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | PLEKHA1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825185
|
Proteintech
26830-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815185
|
Proteintech
26830-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Binds specifically to phosphatidylinositol 3,4-diphosphate (PtdIns3,4P2), but not to other phosphoinositides. May recruit other proteins to the plasma membrane.Specifications
PLEKHA1 | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9HB21 | |
59338 | |
PLEKHA1 Fusion Protein Ag25285 | |
Primary | |
-20°C | |
PLEKHA1 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PLEKHA1, TAPP 1, TAPP1 | |
PLEKHA1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |