Läs mer
Plerixafor, TRC
Produktkod. 17901455
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17901455 50 mg
17921625 100 mg
17911525 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17901455 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP58050050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 110078-46-1
Molekylformel C28 H54 N8
Molekylvikt (g/mol) 502.78
Synonym 1,4,8,11-Tetraazacyclotetradecane, 1,1'-[1,4-phenylenebis(methylene)]bis-, 1,1'-[1,4-Phenylenebis(methylene)]bis[1,4,8,11-tetraazacyclotetradecane], 1,1'-Xylyl bis-1.4,8,11 tetraaza cyclotetradecane, AMD 3100, JKL 169, JM 3100, Mozobil, Plerixafor, SDZ SID 791, 1,4,8,11-Tetraazacyclotetradecane, 1,1'-[1,4-phenylenebis(methylene)]bis-, 1,1'-[1,4-Phenylenebis(methylene)]bis[1,4,8,11-tetraazacyclotetradecane], 1,1'-Xylyl bis-1.4,8,11 tetraaza cyclotetradecane, AMD 3100, JKL 169, JM 3100, Mozobil, Plerixafor, SDZ SID 791, SID791
IUPAC-namn 1-[[4-(1,4,8,11-tetrazacyclotetradec-1-ylmethyl)phenyl]methyl]-1,4,8,11-tetrazacyclotetradecane
LEDER C(N1CCCNCCNCCCNCC1)c2ccc(CN3CCCNCCNCCCNCC3)cc2
Specifikationer
Kemiskt namn eller material Plerixafor
Kvantitet 50 mg
InChI formel InChI=1S/C28H54N8/c1-9-29-15-17-31-13-3-21-35(23-19-33-11-1)25-27-5-7-28(8-6-27)26-36-22-4-14-32-18-16-30-10-2-12-34-20-24-36/h5-8,29-34H,1-4,9-26H2
Formel vikt 502.4471