Läs mer
Invitrogen™ Plexin-A1 Recombinant Rabbit Monoclonal Antibody (JU50-37)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532804
Beskrivning
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Coreceptor for SEMA3A, SEMA3C, SEMA3F and SEMA6D. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm. [UniProt].
Specifikationer
| Plexin-A1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P70206, Q9UIW2 | |
| PLXNA1 | |
| Recombinant protein within Human Plexin A1 aa 100-200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot, Western Blot | |
| JU50-37 | |
| Unconjugated | |
| PLXNA1 | |
| 2600013D04Rik; Kiaa4053; mKIAA4053; NOV; NOVP; Plex 1; PlexA1; plexin 1; plexin A1; plexin-1; plexin-A1; PLXN1; Plxna1; RGD1560871; semaphorin receptor NOV | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18844, 362398, 5361 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.