missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Plexin B1 Antibody (559830), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB37491I
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Plexin B1 Monoclonal antibody specifically detects Plexin B1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifikationer
| Plexin B1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5364 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 559830 | |
| Flow Cytometry, CyTOF-ready | |
| plexin B1, SEP | |
| NS0 mouse myeloma cell line transfected with human Plexin B1, Accession # O43157 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction